Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK3/MEK3 Rabbit anti-Human, Mouse, Rat, Clone: 8U2X3, Novus Biologicals™
SDP

Catalog No. NB167417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167417 100 μg
NB167418 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167417 Supplier Novus Biologicals Supplier No. NBP316163100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MKK3/MEK3 Monoclonal antibody specifically detects MKK3/MEK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen MKK3/MEK3
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 8U2X3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias MAP kinase kinase 3, MAPK/ERK kinase 3, MAPKK 3, MAPKK3PRKMK3dual specificity mitogen-activated protein kinase kinase 3, MEK3MEK 3, mitogen activated protein kinase kinase 3, mitogen-activated protein kinase kinase 3, MKK3EC 2.7.12.2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-347 of human MKK3/MEK3 (P46734).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Hypoxia, Lipid and Metabolism, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5606
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.