Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKK3/MEK3 Rabbit anti-Human, Mouse, Rat, Clone: 8U2X3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | MKK3/MEK3 |
---|---|
Clone | 8U2X3 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
MKK3/MEK3 Monoclonal antibody specifically detects MKK3/MEK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
MKK3/MEK3 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Hypoxia, Lipid and Metabolism, Protein Kinase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5606 | |
IgG | |
Affinity purified |
8U2X3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
MAP kinase kinase 3, MAPK/ERK kinase 3, MAPKK 3, MAPKK3PRKMK3dual specificity mitogen-activated protein kinase kinase 3, MEK3MEK 3, mitogen activated protein kinase kinase 3, mitogen-activated protein kinase kinase 3, MKK3EC 2.7.12.2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 150-347 of human MKK3/MEK3 (P46734). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title