Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK4/MEK4 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB084755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084755 Supplier Novus Biologicals Supplier No. NBP299331B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MKK4/MEK4 Polyclonal specifically detects MKK4/MEK4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MKK4/MEK4
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias c-Jun N-terminal kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 4, JNK activating kinase 1, JNK-activated kinase 1, JNK-activating kinase 1, JNKK, JNKK1MAPK/ERK kinase 4, MAP kinase kinase 4, MAPKK4, MEK4MEK 4, mitogen-activated protein kinase kinase 4, MKK4SAPK/ERK kinase 1, PRKMK4MAPKK 4, SEK1, SERK1EC 2.7.12.2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human MKK4/MEK4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6416
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.