Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK7/MEK7, p Thr277 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229678 0.1 mL
NB019425 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229678 Supplier Novus Biologicals Supplier No. NBP229678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MKK7/MEK7 Polyclonal specifically detects MKK7/MEK7 in Human samples. It is validated for Western Blot.

Specifications

Antigen MKK7/MEK7 (p Thr277)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation 0.1M Tris-Glycine (phH 7.4), 0.15M NaCl and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. O14733
Gene Alias c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKK7MAPKK 7, PRKMK7
Gene Symbols MAP2K7
Host Species Rabbit
Immunogen peptide corresponding to amino acids surrounding Thr275/Ser277 of human MKK7/SKK4.
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5609
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.