Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKK7/MEK7, p Thr277 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $670.50
Specifications
Antigen | MKK7/MEK7 (p Thr277) |
---|---|
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MKK7/MEK7 Polyclonal specifically detects MKK7/MEK7 in Human samples. It is validated for Western Blot.Specifications
MKK7/MEK7 (p Thr277) | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
O14733 | |
5609 | |
peptide corresponding to amino acids surrounding Thr275/Ser277 of human MKK7/SKK4. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
0.1M Tris-Glycine (phH 7.4), 0.15M NaCl and 30% Glycerol with 0.05% Sodium Azide | |
c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKK7MAPKK 7, PRKMK7 | |
MAP2K7 | |
IgG | |
Affinity Purified | |
47 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title