Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mkl1 Antibody (OTI3G7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272715A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272715A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272715A48 Supplier Novus Biologicals Supplier No. NBP272715AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mkl1 Monoclonal specifically detects Mkl1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Mkl1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G7
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AMKL, KIAA1438, MALSAP and coiled-coil domain, megakaryoblastic leukemia (translocation) 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, MKL/myocardin-like protein 1, Myocardin-related transcription factor A, RNA-binding motif protein 15/megakaryoblastic leukemia-1 fusion protein
Gene Symbols MRTFA
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 165-397 of human Mkl1 (NP_065882) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57591.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.