Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKLP1 Rabbit anti-Human, Rat, Clone: 9V4Y8, Novus Biologicals™
SDP

Catalog No. NB168105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168105 100 μg
NB168106 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168105 Supplier Novus Biologicals Supplier No. NBP316507100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MKLP1 Monoclonal antibody specifically detects MKLP1 in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen MKLP1
Applications Western Blot
Classification Monoclonal
Clone 9V4Y8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias kinesin family member 23, kinesin-like 5 (mitotic kinesin-like protein 1), Kinesin-like protein 5, kinesin-like protein KIF23, KNSL5CHO1, mitotic kinesin-like 1, MKLP-1, MKLP1Mitotic kinesin-like protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 801-960 of human MKLP1 (Q02241).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9493
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.