Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKP-3/DUSP6 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB12019115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12019115 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12019115 Supplier Novus Biologicals Supplier No. NB12019115
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

MKP-3/DUSP6 Polyclonal specifically detects MKP-3/DUSP6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DUSP6a
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:200
Formulation PBS (pH 7.2) with 0.02% Sodium Azide
Gene Alias dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1dual specificity protein phosphatase 6
Gene Symbols DUSP6
Host Species Chicken
Immunogen Synthetic peptide, corresponding to amino acids 135-280 of Human DUSP6a.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1848
Test Specificity DUSP6a
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.