Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKRN1 Antibody (OTI3F9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP272716V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272716V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272716V Supplier Novus Biologicals Supplier No. NBP272716V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MKRN1 Monoclonal specifically detects MKRN1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen MKRN1
Applications Western Blot
Classification Monoclonal
Clone OTI3F9
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias E3 ubiquitin-protein ligase makorin-1, EC 6.3.2.-, FLJ21334, makorin ring finger protein 1, RNF61RING finger protein 61
Gene Symbols MKRN1
Host Species Mouse
Immunogen Full length recombinant protein of human MKRN1 (NP_038474) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 23608.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.