Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKRN2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291912 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291912 Supplier Novus Biologicals Supplier No. NB10055250PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MKRN2 Polyclonal antibody specifically detects MKRN2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen MKRN2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 6.3.2.-, HSPC070, makorin ring finger protein 2, probable E3 ubiquitin-protein ligase makorin-2, RNF62RING finger protein 62
Host Species Rabbit
Immunogen Maps to a region between residue 366 and the C-terminus (residue 416) of human makorin, ring finger protein, 2 using the numbering given in entry NP_054879.3 (GeneID 23609).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 23609
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.