Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKRN3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341688 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341688 Supplier Novus Biologicals Supplier No. NBP281964PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MKRN3 Polyclonal antibody specifically detects MKRN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MKRN3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 6.3.2.-, makorin ring finger protein 3, MGC88288, RNF63D15S9, ZFP127RING finger protein 63, Zinc finger protein 127, ZNF127probable E3 ubiquitin-protein ligase makorin-3
Host Species Rabbit
Immunogen MKRN3 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MKRN3. The immunogen is located within amino acids 400 - 450 of MKRN3. Amino Acid Squence: FYKHEYPEGWGDEPPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7681
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.