Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MKT 077

Click to view available options
Quantity:
50 mg
Description
Occupies mortalin-2 (mot-2), a member of the Hsp70 family, at its p53 binding site and enables p53 translocation to the nucleus. Selectively cytotoxic; causes growth arrest of cancer cells in culture. Also inhibits telomerase activity and cross-links F-actin.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 432 |
Purity | >98% |
Molecular Weight (g/mol) | 432.00 |
Chemical Name or Material | MKT 077 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction