Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 133 hydrochloride
GREENER_CHOICE

Catalog No. p-4817144
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Kir2 channel blocker

Selective blocker of inwardly rectifying Kir2 potassium channels (IC50 values are 1.8, 2.8, 2.9 and 4.0 μM at pH 7.4 for mKir2.1, hKir2.6, hKir2.2 and hKir2.3 respectively). Exhibits no effect on rKir1.1 (IC50 > 300 μM); displays weak activity at hKir7.1 and rKir4.1 (IC50 values are 33 and 76 μM).

Chemical Identifiers

CAS 1222781-70-5
Molecular Formula C19H20ClNO

Specifications

Quantity 10 mg
Formula Weight 313.82
Purity >99%
Molecular Weight (g/mol) 313.83
Chemical Name or Material ML 133 hydrochloride
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.