Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 162

Click to view available options
Quantity:
10 mg
50 mg
Description
ML 162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor. It induces ferroptosis in vivo. It is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50 = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).
Specifications
Specifications
Chemical Name or Material | 2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide |
CAS | 1035072-16-2 |
Quantity | 50 mg |
Target | Glutathione Peroxidase 4/GPX4 |
Molecular Formula | C23H22Cl2N2O3S |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction