Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 190

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective κ opioid receptor (KOP) antagonist (IC50 = 120 nM in a β-arrestin assay); displays >267-fold selectivity over μ and δ opioid receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 488.58 |
| Molecular Weight (g/mol) | 488.592 |
| Chemical Name or Material | ML 190 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction