Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 218 hydrochloride
GREENER_CHOICESDP

Catalog No. 450750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-075-0 50 mg
45-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-075-0 Supplier R&D Systems™ Supplier No. 4507/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CaV3.x blocker

ML 218 hydrochloride is a selective inhibitor of T-type calcium channels (IC50 values are 270 and 310 nM for Cav3.3 and Cav3.2 respectively in a patch EP assay). Decreases burst activity in STN neurons; reduces cataleptic behaviour in an in vivo rat model of Parkinson's disease. Displays no significant inhibition of L- or N-type calcium channels, Kir6 (KATP) or KV11.1 (hERG) potassium channels. ML 218 hydrochloride inhibits oral cancer cell proliferation. Orally active.

Chemical Identifiers

CAS 2319922-08-0
Molecular Formula C19H27Cl3N2O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 405.79
Purity >99%
Molecular Weight (g/mol) 405.788
Chemical Name or Material ML 218 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.