Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 221
GREENER_CHOICE

Catalog No. 474810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-481-0 10 mg
47-485-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-481-0 Supplier Tocris Bioscience™ Supplier No. 4748/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Apelin antagonist

Apelin receptor (APJ) antagonist (IC50 values are 0.70 and 1.75 μM in a cAMP assay and β-arrestin assay, respectively). Displays >37-fold selectivity over the closely related angiotensin II type 1 (AT1) receptor. Exhibits no toxicity towards human hepatocytes at concentrations >50 μM.

Chemical Identifiers

CAS 877636-42-5
Molecular Formula C17H11N3O6S

Specifications

Quantity 10 mg
Formula Weight 385.35
Purity >99%
Molecular Weight (g/mol) 385.35
Chemical Name or Material ML 221
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.