Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Tocris Bioscience™ ML 240
 
    
    
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                10 mg
                            
                        
                            
                                50 mg
                            
                        
                    Description
ML 240 is an ATP-competitive inhibitor of p97 ATPase (VCP, IC50 = 110 nM). Exhibits antiproliferative activity in NCI-60 cancer cell lines and rapidly induces executioner caspases 3 and 7 in multiple colon cancer cells. Promotes accumulation of LC3-II and impairs autophagosome maturation. Also impairs the endoplasmic-reticulum-associated degradation (ERAD) pathway.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| Quantity | 10 mg | 
| Product Type | ML 240 | 
| Purity | >99% | 
| Molecular Weight (g/mol) | 396.44 | 
| Inhibitors | ML 240 | 
For Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                