Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 252

Click to view available options
Quantity:
10 mg
50 mg
Description
ML 252 is a potent and selective inhibitor of KV7.2 voltage-gated potassium channels (IC50 = 70 nM), which displays >40-fold selectivity for KV7.2 over KV7.1 channels (IC50 = 2.9 μM for KV7.1). ML 252 is the (S)-enantiomer, and is more potent than the (R)-enantiomer or racemic mixture (IC50 values are 944 and 160 nM, respectively). ML 252 displays selectivity for KV7.2 over a panel of >68 GPCRs, ion channels and transporters; inhibits melatonin MT1 receptor 60% at 10 μM. ML 252 is brain penetrant.
Specifications
Specifications
Chemical Name or Material | (S)-2-Phenyl-N-(2-(pyrrolidin-1-yl)phenyl)butanamide |
CAS | 1392494-64-2 |
Quantity | 10 mg |
Target | Voltage-gated Potassium Channel Blockers |
Molecular Formula | C20H24N2O |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction