Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ML 311
Click to view available options
Quantity:
10 mg
50 mg
Description
ML 311 is an inhibits Mcl1-BIM interaction. Inhibits viability of Mcl1-dependent tumor cell lines. Induces cell death and decreased spheroid formation in Her2 positive breast cancer cells.
Specifications
Specifications
Chemical Name or Material | 7-[(4-Ethyl-1-piperazinyl)[4-(trifluoromethyl)phenyl]methyl]-8-quinolinol |
CAS | 315698-17-0 |
Assay Percent Range | >99% |
Quantity | 10 mg |
Molecular Formula | C23H24F3N3O |
Formula Weight | Observed MW: 415.45 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction