Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 311

Catalog No. p-7116082
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits Mcl-1-BIM interaction

ML 311 is an inhibits Mcl1-BIM interaction. Inhibits viability of Mcl1-dependent tumor cell lines. Induces cell death and decreased spheroid formation in Her2 positive breast cancer cells.

Specifications

Chemical Name or Material 7-[(4-Ethyl-1-piperazinyl)[4-(trifluoromethyl)phenyl]methyl]-8-quinolinol
CAS 315698-17-0
Assay Percent Range >99%
Quantity 10 mg
Molecular Formula C23H24F3N3O
Formula Weight Observed MW: 415.45
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.