Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 335
GREENER_CHOICE

Catalog No. p-7166813
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective K2P2.1 (TREK-1) and K2P10.1 (TREK-2) activator

ML 335 is a selective K2P2.1 (TREK-1; KCNK2) and K2P10.1 (TREK-2; KCNK10) activator (EC50 values of 14.3 μM and 5.2 μM, respectively); directly stimulates the C-type gate via the K2P modulator pocket. Does not activate K2P4.1 (TRAAK).

Specifications

Quantity 10 mg
Molecular Formula C15H14Cl2N2O3S
Formula Weight 373.25
Product Type Activator
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.