Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 337
GREENER_CHOICE

Catalog No. p-5545889
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective negative allosteric modulator of mGlu3

ML 337 is a selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.

Specifications

Quantity 10 mg
Product Type ML 337
Purity >98%
Molecular Weight (g/mol) 353.39
Inhibitors ML 337

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.