Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 339
GREENER_CHOICE

Catalog No. p-7121016
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 59-431-0

Supplier: Tocris Bioscience™ 5943/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective hCXCR6 antagonist

ML 339 is a potent and selective hCXCR6 antagonist (IC50 = 140 nM); 100-fold less active at the murine CXCR6 receptor (IC50 = 18 μM). Exhibits selectivity over CXCR5, CXCR4, CCR6 and APJ receptors (IC50 >79 μM).

Specifications

Chemical Name or Material N-[(1 R,3 S,5 S)-9-(2-((2-Chlorophenyl)amino)-2-exoethyl)-9-azabicyclo[3.3.1 ]nonan-3-yl)-3,4,5-trimethoxybenzamide
CAS 2080300-49-6
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Target Chemokine Receptor Antagonists
Molecular Formula C26H32ClN3O5
Formula Weight 502
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity 98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.