Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 349
Click to view available options
Quantity:
10 mg
50 mg
Description
ML 349 is a selective and reversible lysophospholipase 2 (LYPLA2) inhibitor (IC50 = 144 nM). Displays selectively over LYPLA1 and 20 other serine hydrolases (IC50 values are >3000 and >10,000 nM respectively).
Specifications
Specifications
Purity | >98% |
Quantity | 50 mg |
Product Type | ML 349 |
Molecular Weight (g/mol) | 454.56 |
Inhibitors | ML 349 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction