Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 351

Catalog No. p-7146018
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 12/15 LOX inhibitor; active in vivo

ML 351 is a selective 12/15 LOX inhibitor (IC50 = 200 nM). Exhibits >250-fold selectivity over related isozymes, 5-LOX, platelet 12-LOX, 15-LOX-2, ovine COX-1, and human COX-2. Protects against oxidative glutamate toxicity in mouse neuronal cells (HT-22) and reduces infarct size in a mouse ischemic stroke model.

Specifications

Chemical Name or Material 5-(Methylamino)-2-(1-naphthalenyl)-4-oxazolecarbonitrile
CAS 847163-28-4
Quantity 5 mg
Molecular Formula C15H11N3O
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.