Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 418

Click to view available options
Quantity:
10 mg
50 mg
Description
ML 418 is a Kir7.1 inhibitor (IC50 = 0.31 μM). Also inhibits Kir6.2 with similar potency. Displays >17-fold selectivity for Kir7.1 over Kir1.1, Kir2.1, Kir2.2, Kir2.3, Kir3.1/3.2 and Kir4.1. Active in vivo.
Specifications
Specifications
Chemical Name or Material | Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate |
CAS | 1928763-08-9 |
Quantity | 10 mg |
Molecular Formula | C19H24CIN3O3 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction