Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML SA1
GREENER_CHOICE

Catalog No. 474610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-461-0 10 mg
47-465-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-461-0 Supplier Tocris Bioscience™ Supplier No. 4746/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Activator of TRPML channels

Activator of TRPML channels (TRPML1, 2 and 3); does not activate TRPM2, TRPV2, TRPV3, TRPC6 or TRPA1 channels. Induces TRPML-mediated Ca2+ release from lysosomes; activity corrects trafficking defects and reduces cholesterol accumulation in Niemann-Pick type C macrophages.

Chemical Identifiers

CAS 332382-54-4
Molecular Formula C22H22N2O3

Specifications

Quantity 10 mg
Formula Weight 362.42
Purity >99%
Molecular Weight (g/mol) 362.429
Chemical Name or Material ML SA1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.