Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML SA1
GREENER_CHOICE

Catalog No. p-4817007
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Activator of TRPML channels

Activator of TRPML channels (TRPML1, 2 and 3); does not activate TRPM2, TRPV2, TRPV3, TRPC6 or TRPA1 channels. Induces TRPML-mediated Ca2+ release from lysosomes; activity corrects trafficking defects and reduces cholesterol accumulation in Niemann-Pick type C macrophages.

Chemical Identifiers

CAS 332382-54-4
Molecular Formula C22H22N2O3

Specifications

Quantity 10 mg
Formula Weight 362.42
Purity >99%
Molecular Weight (g/mol) 362.429
Chemical Name or Material ML SA1
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.