Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLC1SA Antibody, Novus Biologicals™
SDP

Catalog No. NBP17424720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17424720 20 μL
NBP174247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17424720 Supplier Novus Biologicals Supplier No. NBP17424720UL

Rabbit Polyclonal Antibody

MLC1SA Polyclonal specifically detects MLC1SA in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen LOC100359980
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. D3ZHA7
Gene Alias smooth muscle and non-muscle myosin alkali light chain 6B-like
Gene Symbols LOC100359980
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the N terminal of LOC100359980. Immunizing peptide sequence: PKKDAPVKKPAGPSISKPAAKSTAGTPLAKTKAEPAAPQTPVKTQEPPVD.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100359980
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.