Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLC1SA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | LOC100359980 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17424720
![]() |
Novus Biologicals
NBP17424720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174247
![]() |
Novus Biologicals
NBP174247 |
100 μL |
Each for $487.50
|
|
|||||
Description
MLC1SA Polyclonal specifically detects MLC1SA in Rat samples. It is validated for Western Blot.Specifications
LOC100359980 | |
Polyclonal | |
Rabbit | |
D3ZHA7 | |
100359980 | |
IgG | |
23 kDa |
Western Blot | |
Unconjugated | |
RUO | |
smooth muscle and non-muscle myosin alkali light chain 6B-like | |
LOC100359980 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title