Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLC1SA Antibody, Novus Biologicals™
SDP

Catalog No. NBP217304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217304 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217304 Supplier Novus Biologicals Supplier No. NBP217304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLC1SA Polyclonal specifically detects MLC1SA in Human samples. It is validated for Western Blot.

Specifications

Antigen MLC1SA
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P14649
Gene Alias MLC1sa, MLC1SAmyosin light chain 1, slow-twitch muscle A isoform, myosin alkali light chain 1 slow a, myosin light chain 1 slow a, Myosin light chain 1 slow-twitch muscle A isoform, myosin light chain 6B, myosin, light chain 6B, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle, smooth muscle and non-muscle myosin alkali light chain 6B, Smooth muscle and nonmuscle myosin light chain alkali 6B
Gene Symbols MYL6B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MLC1SA. The exact sequence is proprietary.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140465
Test Specificity Human. Species reactivity based on sequence identity: Mouse (96%), Bovine (88%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.