Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLC1SA Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB326801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326801 Supplier Novus Biologicals Supplier No. NBP299163PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLC1SA Polyclonal antibody specifically detects MLC1SA in Human samples. It is validated for Western Blot

Specifications

Antigen MLC1SA
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias MLC1sa, MLC1SAmyosin light chain 1, slow-twitch muscle A isoform, myosin alkali light chain 1 slow a, myosin light chain 1 slow a, Myosin light chain 1 slow-twitch muscle A isoform, myosin light chain 6B, myosin, light chain 6B, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle, smooth muscle and non-muscle myosin alkali light chain 6B, Smooth muscle and nonmuscle myosin light chain alkali 6B
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MLC1SA
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140465
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.