Learn More
Description
Specifications
Specifications
| Antigen | MLF1 Interacting Protein (p Thr78) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:25000, Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Dot Blot |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | NP_078905 |
| Gene Alias | CENP-50, CENP-U, CENPU50, CENPUMLF1-interacting protein, centromere protein of 50 kDa, centromere protein U, FLJ23468, ICEN24, Interphase centromere complex protein 24, KLIP1CENP50, KSHV latent nuclear antigen interacting protein 1, KSHV latent nuclear antigen-interacting protein 1, MLF1 interacting protein, PBIP1, Polo-box-interacting protein 1 |
| Gene Symbols | CENPU |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
