Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLF1 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
SDP

Catalog No. 501675383
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-5383 100 μL
50-254-0129 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-5383 Supplier OriGene Supplier No. TA504765
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLF1 is involved in lineage commitment of primary hemopoietic progenitors by restricting erythroid formation and enhancing myeloid formation. The protein interferes with erythopoietin-induced erythroid terminal differentiation by preventing cells from exiting the cell cycle through suppression of CDKN1B/p27Kip1 levels. It suppresses RFWD2/COP1 activity via CSN3 which activates p53 and induces cell cycle arrest. It binds DNA and affects the expression of a number of genes so may function as a transcription factor in the nucleus.

Specifications

Antigen MLF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MLF1
Gene Accession No. P58340
Gene Alias myeloid leukemia factor 1
Gene Symbols MLF1
Host Species Mouse
Immunogen Full length human recombit protein of human MLF1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4291
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.