Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLH1 Antibody (164C819), Novus Biologicals™
SDP

Catalog No. NB10056552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056552 0.1 mg
NB10056552T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056552 Supplier Novus Biologicals Supplier No. NB10056552
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLH1 Monoclonal specifically detects MLH1 in Human samples. It is validated for Western Blot.

Specifications

Antigen MLH1
Applications Western Blot
Classification Monoclonal
Clone 164C819
Conjugate Unconjugated
Dilution Western Blot 2-3 ug/ml
Gene Accession No. P40692
Gene Alias COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1
Gene Symbols MLH1
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 387-403 DSREQKLDAFLQPLSKP of human MLH1; GenBank no. NP_000240.1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Repair, Mismatch Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4292
Test Specificity In Molt-4, a 83 kDa band is observed.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.