Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLH1 Antibody (MLH1/6284R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB155302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155302 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB155302 Supplier Novus Biologicals Supplier No. NBP314124JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MLH1 Monoclonal antibody specifically detects MLH1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MLH1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MLH1/6284R
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1
Host Species Rabbit
Immunogen Recombinant full-length human MLH1 protein (Uniprot: P40692)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4292
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.