Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLH1 Mouse anti-Human, Clone: G168-728, Novus Biologicals™
SDP

Catalog No. NB122114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122114 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122114 Supplier Novus Biologicals Supplier No. NBP30732020UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLH1 Monoclonal specifically detects MLH1 in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen MLH1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G168-728
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 2-4 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1
Host Species Mouse
Immunogen Recombinant full-length human MLH1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4292
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.