Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLH1 Mouse anti-Human, Clone: MLH1/1324, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | MLH1 |
|---|---|
| Clone | MLH1/1324 |
| Dilution | Western Blot 0.5-1.0ug/ml, SDS-Page |
| Applications | Western Blot, SDS-Page |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247656A
![]() |
Novus Biologicals
NBP2476560.1MG |
0.1 mg | N/A | N/A | N/A | ||||
NBP247656B
![]() |
Novus Biologicals
NBP2476560.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
MLH1 Monoclonal specifically detects MLH1 in Human samples. It is validated for ELISA.Specifications
| MLH1 | |
| Western Blot 0.5-1.0ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1 | |
| MLH1 | |
| IgG2b | |
| Protein A or G purified | |
| This MAb recognizes a protein of 83kDa, identified as MLH1. Its epitope maps in aa 380-410. Defects in MLH1 are the cause of hereditary non-polyposis colorectal cancer type 2 (HNPCC2). Heterodimerizes with PMS2 to form MutL alpha, a component of the post-replicative DNA mismatch repair system (MMR). DNA repair is initiated by MutS alpha (MSH2-MSH6) or MutS beta (MSH2-MSH6) binding to a dsDNA mismatch, then MutL alpha is recruited to the heteroduplex. Assembly of the MutL-MutS-heteroduplex ternary complex in presence of RFC and PCNA is sufficient to activate endonuclease activity of PMS2. It introduces single-strand breaks near the mismatch and thus generates new entry points for the exonuclease EXO1 to degrade the strand containing the mismatch. DNA methylation would prevent cleavage and therefore assure that only the newly mutated DNA strand is going to be corrected. MutL alpha (MLH1-PMS2) interacts physically with the clamp loader subunits of DNA polymerase III, suggesting that it may play a role to recruit the DNA polymerase III to the site of the MMR. Also implicated in DNA damage signaling, a process, which induces cell cycle arrest and can lead to apoptosis in case of major DNA damages. Heterodimerizes with MLH3 to form MutL gamma, which plays a role in meiosis. |
| MLH1/1324 | |
| Western Blot, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 4292 | |
| Recombinant human MLH1 protein | |
| Primary | |
| Store at 4C. | |
| 85 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title