Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLK2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB132315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132315 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132315 Supplier Novus Biologicals Supplier No. NBP306104AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLK2 Polyclonal specifically detects MLK2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MLK2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, MEKK10, mitogen-activated protein kinase kinase kinase 10, Mixed lineage kinase 2, MKN28 kinase, MLK2MKN28 derived nonreceptor_type serine/threonine kinase, MSTEC 2.7.11.25, Protein kinase MST
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MLK2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 4294
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.