Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1457110.025MG
Description
MLK3 Polyclonal specifically detects MLK3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MLK3 | |
Polyclonal | |
Unconjugated | |
PBS, 1% BSA, 50% Glycerol with 0.02% Thimerosal | |
EC 2.7.11, EC 2.7.11.25, MEKK11, mitogen-activated protein kinase kinase kinase 11, Mixed lineage kinase 3, MLK3MLK-3, PTK1protein-tyrosine kinase PTK1, SH3 domain-containing proline-rich kinase, SPRKMGC17114, Src-homology 3 domain-containing proline-rich kinase | |
Rabbit | |
93 kDa | |
0.025 mg | |
Protein Kinase | |
4296 | |
Human, Rat, Mouse | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 0.5 - 4 ug/mL | |
Q16584 | |
MAP3K11 | |
Synthetic peptide mapping to the C-terminus of human MLK3. | |
Affinity Purified | |
RUO | |
Primary | |
It does not cross-react with MLK1, MLK2 and other mixed lineage kinases. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction