Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$220.00 - $524.00
Specifications
Antigen | MLK3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
MLK3 Polyclonal specifically detects MLK3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MLK3 | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
PBS, 1% BSA, 50% Glycerol with 0.02% Thimerosal | |
EC 2.7.11, EC 2.7.11.25, MEKK11, mitogen-activated protein kinase kinase kinase 11, Mixed lineage kinase 3, MLK3MLK-3, PTK1protein-tyrosine kinase PTK1, SH3 domain-containing proline-rich kinase, SPRKMGC17114, Src-homology 3 domain-containing proline-rich kinase | |
MAP3K11 | |
IgG | |
Affinity Purified | |
It does not cross-react with MLK1, MLK2 and other mixed lineage kinases. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat, Mouse | |
Q16584 | |
4296 | |
Synthetic peptide mapping to the C-terminus of human MLK3. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title