Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLKL, p Thr357 Antibody (954724), HRP, R&D Systems™
SDP

Catalog No. MAB91871H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB91871H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB91871H Supplier Novus Biologicals Supplier No. MAB91871H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLKL Monoclonal specifically detects MLKL in Human samples. It is validated for Flow Cytometry (Negative), Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MLKL
Applications Flow Cytometry (Negative), Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 954724
Conjugate HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow Cytometry (Negative), Immunocytochemistry
Formulation PBS with No Preservative
Gene Alias FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein
Gene Symbols MLKL
Host Species Mouse
Immunogen Synthetic peptide from human MLKL phosphorylated at T357 [UniProt Q8NB16]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 197259
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.