Learn More
Description
Specifications
Specifications
| Antigen | MLKL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of mouse MLKL. |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
