Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLKL Rabbit anti-Human, Mouse, Rat, Clone: 7L1P5, Novus Biologicals™
SDP

Catalog No. NB166813 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166813 100 μg
NB166814 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166813 Supplier Novus Biologicals Supplier No. NBP315859100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MLKL Monoclonal antibody specifically detects MLKL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MLKL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7L1P5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-193 of human MLKL (Q8NB16).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Necroptosis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 197259
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.