Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLKL Rabbit anti-Human, Mouse, Rat, Clone: 7L1P5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $504.00
Specifications
Antigen | MLKL |
---|---|
Clone | 7L1P5 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
MLKL Monoclonal antibody specifically detects MLKL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MLKL | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 10-193 of human MLKL (Q8NB16). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7L1P5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Necroptosis, Protein Kinase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
197259 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title