Learn More
MLL1 Recombinant Rabbit Monoclonal Antibody (BL-175-7E8), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $378.84
Specifications
Antigen | MLL1 |
---|---|
Clone | BL-175-7E8 |
Applications | Immunoprecipitation, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1274
![]() |
Bethyl Laboratories, Inc
A700010T |
20 μL |
Each for $157.08
|
|
|||||
50-157-1273
![]() |
Bethyl Laboratories, Inc
A700010 |
100 μL |
Each for $378.84
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.Specifications
MLL1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q03164 | |
4297 | |
Between 720 and 780 | |
Primary | |
4° C | |
KMT2A |
BL-175-7E8 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
ALL-1; Trithorax-like protein | |
KMT2A | |
IgG | |
Ion-exchange chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.