Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25773025UL
Description
MLL3 Polyclonal specifically detects MLL3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MLL3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
ALR-like protein, DKFZp686C08112, EC 2.1.1.43, FLJ12625, HALRMGC119851, histone-lysine N-methyltransferase MLL3, histone-lysine N-methyltransferase, H3 lysine-4 specific, Homologous to ALR protein, KIAA1506FLJ38309, KMT2C, KMT2CMGC119852, Lysine N-methyltransferase 2C, MGC119853, myeloid/lymphoid or mixed-lineage leukemia 3, Myeloid/lymphoid or mixed-lineage leukemia protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
58508 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
KMT2C | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TPPTMSQPTFPMVPQQLQHQQHTTVISGHTSPVRMPSLPGWQPNSAPAHLPLNPPRIQPPIAQLPIKTCTPAPGTVSNANPQ | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction