Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLLT6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP217307 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217307 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217307 Supplier Novus Biologicals Supplier No. NBP217307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLLT6 Polyclonal specifically detects MLLT6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MLLT6
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.32mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Alias AF17Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6, ALL1-fused gene from chromosome 17 protein, FLJ23480, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 6, protein AF-17, trithorax homolog
Gene Symbols MLLT6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MLLT6. The exact sequence is proprietary.
Molecular Weight of Antigen 112 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4302
Test Specificity Human. Species reactivity based on sequence identity: mouse (92%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.