Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MLN 4924

Click to view available options
Quantity:
10 mg
50 mg
Description
MLN 4924 (Tak 924) is a potent and selective NEDD8 activating enzyme (NAE) inhibitor (IC50 = 4.7 nM). MLN 4924 exhibits selectivity over closely related enzymes UAE, UBA6, SAE, and ATG7 (IC50 = 1.5, 1.8, 8.2, and >10 μM, respectively), and displays minimal activity at adenosine receptors and a panel of 12 cellular kinases. MLN 4924 exhibits cyotoxicity in several human tumor-derived cell lines and causes liver damage in SD rats, when given with TNF-α. It blocks the NEDD8 pathway causing DNA damage in mice bearing HCT-116 xenografts. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | [(1 S,2 S,4 R)-4-[4-[[(1 S)-2,3-Dihydro-1 H-inden-1-yl]amino]-7 H-pyrrolo[2,3-d]pyrimidin-7-yl]-2-hydroxycyclopentyl]methyl sulfamic acid ester |
| CAS | 905579-51-3 |
| Quantity | 10 mg |
| Target | NEDD8 Activating Enzyme E1) |
| Molecular Formula | C21H25N5O4S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction